accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3536-96-7

IUPAC Name: chloro(ethenyl)magnesium
Synonyms: chloro(ethenyl)magnesium
Molecular Weight (g/mol): 86.8
Molecular Formula: C2H3ClMg
InChi Key: NAFZXKWEUWVSAW-UHFFFAOYSA-M
SMILES: Cl[Mg]C=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)