accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35370-95-7

IUPAC Name: 2-(3-chlorophenoxy)ethanethioamide
Synonyms: 2-(3-chlorophenoxy)ethanethioamide
Molecular Weight (g/mol): 201.67
Molecular Formula: C8H8ClNOS
InChi Key: RPAOLVIADVQKNA-UHFFFAOYSA-N
SMILES: NC(=S)COC1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results