accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClCH3ONHN

CAS RN 35386-24-4

IUPAC Name: dihydrogen 1-(2-methoxyphenyl)piperazine dichloride
Synonyms: dihydrogen 1-(2-methoxyphenyl)piperazine dichloride
Molecular Weight (g/mol): 265.18
Molecular Formula: C11H18Cl2N2O
InChi Key: ZGWQDMTYAQEMHA-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)