accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3540-78-1

IUPAC Name: N-(3,3'-dichloro-4'-{[(4-methoxyphenyl)methylidene]amino}-[1,1'-biphenyl]-4-yl)-1-(4-methoxyphenyl)methanimine
Synonyms: N-(3,3'-dichloro-4'-{[(4-methoxyphenyl)methylidene]amino}-[1,1'-biphenyl]-4-yl)-1-(4-methoxyphenyl)methanimine
Molecular Weight (g/mol): 489.4
Molecular Formula: C28H22Cl2N2O2
InChi Key: JWTLKDNURNVSBN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=NC2=C(Cl)C=C(C=C2)C2=CC(Cl)=C(C=C2)N=CC2=CC=C(OC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results