accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3541-42-2

IUPAC Name: (2E)-3-(2-hydroxyphenyl)prop-2-enal
Synonyms: (2E)-3-(2-hydroxyphenyl)prop-2-enal
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: BSDNZCQPDVTDET-HWKANZROSA-N
SMILES: OC1=CC=CC=C1\C=C\C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results