accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBrBr

CAS RN 354135-75-4

IUPAC Name: 6,6'-dibromo-9,9'-diphenyl-9H,9'H-3,3'-bicarbazole
Synonyms: 6,6'-dibromo-9,9'-diphenyl-3,3'-bicarbazole
Molecular Weight (g/mol): 642.39
Molecular Formula: C36H22Br2N2
InChi Key: VTIPHPNQFUCDAW-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC2=C(C=C1)N(C1=C2C=C(Br)C=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results