accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NHOONNClCl

CAS RN 3543-75-7

IUPAC Name: hydrogen 4-{5-[bis(2-chloroethyl)amino]-1-methyl-1H-1,3-benzodiazol-2-yl}butanoic acid chloride
Synonyms: bendamustine
Molecular Weight (g/mol): 394.72
Molecular Formula: C16H22Cl3N3O2
InChi Key: ZHSKUOZOLHMKEA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1C(CCCC(O)=O)=NC2=CC(=CC=C12)N(CCCl)CCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results