accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSClOO

CAS RN 35432-36-1

IUPAC Name: 2-methylpropane-1-sulfonyl chloride
Synonyms: 2-methylpropane-1-sulfonyl chloride
Molecular Weight (g/mol): 156.62
Molecular Formula: C4H9ClO2S
InChi Key: NQLNHTOCFWUYQE-UHFFFAOYSA-N
SMILES: CC(C)CS(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)