accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOH

CAS RN 35461-98-4

IUPAC Name: 4-(furan-2-yl)benzoic acid
Synonyms: 4-(furan-2-yl)benzoic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: FOJYVBSPOBUCMV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results