accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35466-83-2

IUPAC Name: methyl prop-2-en-1-yl carbonate
Synonyms: methyl prop-2-en-1-yl carbonate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: YHLVIDQQTOMBGN-UHFFFAOYSA-N
SMILES: COC(=O)OCC=C
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)