accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35466-87-6

IUPAC Name: [({2-[(ethoxycarbonyl)oxy]ethoxy}carbonyl)oxy]ethane
Synonyms: [({2-[(ethoxycarbonyl)oxy]ethoxy}carbonyl)oxy]ethane
Molecular Weight (g/mol): 206.19
Molecular Formula: C8H14O6
InChi Key: MYNUAGYBVSQRFN-UHFFFAOYSA-N
SMILES: CCOC(=O)OCCOC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results