accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 354812-17-2

IUPAC Name: 4-amino-[2,3'-bithiophene]-5-carboxamide
Synonyms: 4-amino-[2,3'-bithiophene]-5-carboxamide
Molecular Weight (g/mol): 224.3
Molecular Formula: C9H8N2OS2
InChi Key: BMUACLADCKCNKZ-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N)C=C(S1)C1=CSC=C1
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results