accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 355134-13-3

IUPAC Name: 3-bromo-5-nitrobenzaldehyde
Synonyms: 3-bromo-5-nitrobenzaldehyde
Molecular Weight (g/mol): 230.02
Molecular Formula: C7H4BrNO3
InChi Key: OUDCOMBHRXKPIJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CC(C=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results