accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3OSiOSiCH3H3CCH3OSiH3CCH3H3COSiCH3H3CCH3

CAS RN 3555-47-3

IUPAC Name: tetratrimethylsilyl silicate
Synonyms: tetrakis(trimethylsiloxy)silane
Molecular Weight (g/mol): 384.84
Molecular Formula: C12H36O4Si5
InChi Key: VNRWTCZXQWOWIG-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)