accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNNOCH2Cl

CAS RN 35554-44-0

IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-(prop-2-en-1-yloxy)ethyl]-1H-imidazole
Synonyms: imazalil imazalil base imazalil mononitrate imazalil phosphate imazalil sulfate (1:1) 恩康唑 fungaflor deccozil chloramizol enilconazole
Molecular Weight (g/mol): 297.18
Molecular Formula: C14H14Cl2N2O
InChi Key: PZBPKYOVPCNPJY-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C(CN2C=CN=C2)OCC=C)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results