accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOCH3

CAS RN 3556-60-3

IUPAC Name: 3-methoxy-4-methylbenzonitrile
Synonyms: 3-methoxy-4-methylbenzonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: QLJZMAGXHHXXMS-UHFFFAOYSA-N
SMILES: COC1=C(C)C=CC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results