accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35572-78-2

IUPAC Name: 2-methyl-3,5-dinitroaniline
Synonyms: 2-amino-4,6-dinitrotoluene
Molecular Weight (g/mol): 197.15
Molecular Formula: C7H7N3O4
InChi Key: IEEJAAUSLQCGJH-UHFFFAOYSA-N
SMILES: CC1=C(N)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results