accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35575-96-3

IUPAC Name: dimethyl [({6-chloro-2-oxo-2H,3H-[1,3]oxazolo[4,5-b]pyridin-3-yl}methyl)sulfanyl]phosphonate
Synonyms: azamethiphos ciba-geigy 18809 actogard C9H10ClN2O5PS Snip
Molecular Weight (g/mol): 324.67
Molecular Formula: C9H10ClN2O5PS
InChi Key: VNKBTWQZTQIWDV-UHFFFAOYSA-N
SMILES: COP(=O)(OC)SCN1C(=O)OC2=CC(Cl)=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results