accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 3558-69-8

IUPAC Name: 2,6-diphenylpyridine
Synonyms: 2,6-diphenylpyridine
Molecular Weight (g/mol): 231.3
Molecular Formula: C17H13N
InChi Key: PJUOHDQXFNPPRF-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CC(=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)