accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO(S)OH3CH3CO(R)(S)(S)(S)FO(S)H3C(R)F(S)OH(S)H3C

CAS RN 356-12-7

IUPAC Name: 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-8-yl]-2-oxoethyl acetate
Synonyms: fluocinonide vanos lidex lidex E lidex-E topsyn lyderm fluocinolide topsyne fluzon
Molecular Weight (g/mol): 494.53
Molecular Formula: C26H32F2O7
InChi Key: WJOHZNCJWYWUJD-IUGZLZTKSA-N
SMILES: CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (6)
Percent Purity 
  • (6)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (6)
  • (6)