accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 356-24-1

IUPAC Name: methyl 2,2,3,3,4,4,4-heptafluorobutanoate
Synonyms: methyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular Weight (g/mol): 228.07
Molecular Formula: C5H3F7O2
InChi Key: MRPUVAKBXDBGJQ-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results