accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 356-27-4

IUPAC Name: ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
Synonyms: ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular Weight (g/mol): 242.09
Molecular Formula: C6H5F7O2
InChi Key: JVHJRIQPDBCRRE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results