accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)(R)CH3(S)(R)CH3(R)(S)(S)(R)CH3CH3CH3

CAS RN 35602-69-8

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl octadecanoate
Synonyms: cholesteryl stearate 18:0 cholesteryl ester 5-cholesten-3b-ol stearate cholestryl stearate
Molecular Weight (g/mol): 653.13
Molecular Formula: C45H80O2
InChi Key: XHRPOTDGOASDJS-XNTGVSEISA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results