accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35620-71-4

IUPAC Name: 2-phenoxypyridine-3-carboxylate
Synonyms: 2-phenoxypyridine-3-carboxylate
Molecular Weight (g/mol): 214.2
Molecular Formula: C12H8NO3
InChi Key: CQGAXJGXGLVFGJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CN=C1OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results