accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35620-72-5

IUPAC Name: 2-(phenylsulfanyl)pyridine-3-carboxylate
Synonyms: 2-(phenylsulfanyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 230.26
Molecular Formula: C12H8NO2S
InChi Key: MGIDOIDQQMOYTH-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CN=C1SC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results