accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCH3CH3NHNOSOOOSOOOH3C(E)

CAS RN 3564-09-8

IUPAC Name: disodium (4E)-3-oxo-4-[2-(2,4,5-trimethylphenyl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: ponceau 3R
Molecular Weight (g/mol): 494.44
Molecular Formula: C19H16N2Na2O7S2
InChi Key: IZJTYWDKWZYJNW-GQKUHHGYSA-L
SMILES: [Na+].[Na+].CC1=CC(C)=C(N\N=C2\C(=O)C(=CC3=CC(=CC=C23)S([O-])(=O)=O)S([O-])(=O)=O)C=C1C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)