accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 356561-08-5

IUPAC Name: 5-iodo-4-methylpyridin-2-amine
Synonyms: 5-iodo-4-methylpyridin-2-amine
Molecular Weight (g/mol): 234.04
Molecular Formula: C6H7IN2
InChi Key: HYRZCIXFHXERJT-UHFFFAOYSA-N
SMILES: CC1=CC(N)=NC=C1I
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)