accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35660-94-7

IUPAC Name: (2E)-2-methylbut-2-enoyl chloride
Synonyms: (2E)-2-methylbut-2-enoyl chloride
Molecular Weight (g/mol): 118.56
Molecular Formula: C5H7ClO
InChi Key: TUNLYEHIVPWOHK-ONEGZZNKSA-N
SMILES: C\C=C(/C)C(Cl)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results