accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)(S)CH3OHOH3C(R)(Z)

CAS RN 3569-10-6

IUPAC Name: (2Z)-3-[(4S,7R,7aR)-3,7-dimethyl-2,4,5,6,7,7a-hexahydro-1H-inden-4-yl]-2-methylprop-2-enoic acid
Synonyms: valerenic acid
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: FEBNTWHYQKGEIQ-JWRGBUJQSA-N
SMILES: C[C@@H]1CC[C@@H](\C=C(\C)C(O)=O)C2=C(C)CC[C@H]12
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)