accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHN

CAS RN 3569-21-9

IUPAC Name: 3-(1H-indol-3-yl)propan-1-ol
Synonyms: 3-(1H-indol-3-yl)propan-1-ol
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: LYPSVQXMCZIRGP-UHFFFAOYSA-N
SMILES: OCCCC1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results