accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHNON

CAS RN 3569-99-1

IUPAC Name: N-(hydroxymethyl)pyridine-3-carboxamide
Synonyms: N-(hydroxymethyl)nicotinamide
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: JRFKIOFLCXKVOT-UHFFFAOYSA-N
SMILES: OCNC(=O)C1=CC=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results