CAS RN 35693-99-3
IUPAC Name:
2,2',5,5'-tetrachloro-1,1'-biphenyl
Synonyms:
tetrachlorobiphenyl TCBP
Molecular Weight (g/mol):
291.98
Molecular Formula:
C12H6Cl4
InChi Key:
HCWZEPKLWVAEOV-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(Cl)C(=C1)C1=CC(Cl)=CC=C1Cl
1
–
1
of
1
results
1
Agilent Technologies 2,2',5,5'-Tetrachlorobiphenyl solution, 100μg/mL in hexane, Ultra Scientific
CAS: 35693-99-3 Molecular Formula: C12H6Cl4 Molecular Weight (g/mol): 291.98 InChI Key: HCWZEPKLWVAEOV-UHFFFAOYSA-N PubChem CID: 37248 ChEBI: CHEBI:34206 IUPAC Name: 1,4-dichloro-2-(2,5-dichlorophenyl)benzene SMILES: C1=CC(=C(C=C1Cl)C2=C(C=CC(=C2)Cl)Cl)Cl