accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHgHOOHgOCH3OOHOO

CAS RN 3570-80-7

IUPAC Name: {5'-[(acetyloxy)mercurio]-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-4'-yl}mercurio acetate
Synonyms: 5'-[(acetyloxy)mercurio]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthen]-4'-ylmercurio acetate
Molecular Weight (g/mol): 849.56
Molecular Formula: C24H16Hg2O9
InChi Key: QSZMOZPZBXFCDN-UHFFFAOYSA-L
SMILES: CC(=O)O[Hg]C1=C2OC3=C(C=CC(O)=C3[Hg]OC(C)=O)C3(OC(=O)C4=CC=CC=C34)C2=CC=C1O
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)