accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONOOCH3H3CCH3

CAS RN 357154-16-6

IUPAC Name: 1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate
Molecular Weight (g/mol): 271.31
Molecular Formula: C13H21NO5
InChi Key: YAVQLRUBKDCOCI-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)