accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOONOOOH2OH2OH2OH2OH2OYb

CAS RN 35725-34-9

IUPAC Name: ytterbium(3+) pentahydrate trinitrate
Synonyms: ytterbium(3+) pentahydrate trinitrate
Molecular Weight (g/mol): 449.14
Molecular Formula: H10N3O14Yb
InChi Key: XIOPWXFTXDPBEY-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Yb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (8)
Grade 
  • (8)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results