accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 357437-74-2

CH3NBrBr

CAS RN 357437-74-2

IUPAC Name: 3,6-dibromo-9-(4-methylphenyl)-9H-carbazole
Synonyms: 3,6-dibromo-9-(4-methylphenyl)carbazole
Molecular Weight (g/mol): 415.13
Molecular Formula: C19H13Br2N
InChi Key: ZLCMXHALWTYHOY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)