accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 3575-31-3

IUPAC Name: 4-octylbenzoic acid
Synonyms: benzoic acid, 4-octyl-
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: ZQLDNJKHLQOJGE-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)