accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3575-32-4

IUPAC Name: N1,N1-dimethylbenzene-1,3-diamine dihydrochloride
Synonyms: N1,N1-dimethylbenzene-1,3-diamine dihydrochloride
Molecular Weight (g/mol): 209.11
Molecular Formula: C8H14Cl2N2
InChi Key: BZJPIQKDEGXVFG-UHFFFAOYSA-N
SMILES: Cl.Cl.CN(C)C1=CC=CC(N)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)