accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOO

CAS RN 3577-87-5

IUPAC Name: methyl diphenyl phosphite
Synonyms: methyl diphenyl phosphite
Molecular Weight (g/mol): 248.22
Molecular Formula: C13H13O3P
InChi Key: NYCZNDFWFCCTPA-UHFFFAOYSA-N
SMILES: COP(OC1=CC=CC=C1)OC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results