accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOPOOFFFOFFF

CAS RN 358-63-4

IUPAC Name: tris(2,2,2-trifluoroethyl) phosphate
Synonyms: tris(2,2,2-trifluoroethyl) phosphate
Molecular Weight (g/mol): 344.07
Molecular Formula: C6H6F9O4P
InChi Key: ZMQDTYVODWKHNT-UHFFFAOYSA-N
SMILES: FC(F)(F)COP(=O)(OCC(F)(F)F)OCC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results