accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOONOOHOOHH2ONaNaCd

CAS RN 35803-35-1

IUPAC Name: disodium 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxylatomethyl)amino)acetate cadmium hydrate
Synonyms: disodium 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxylatomethyl)amino)acetate cadmium hydrate
Molecular Weight (g/mol): 466.64
Molecular Formula: C10H16CdN2Na2O9
InChi Key: AZRABGJSTJUALZ-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].[Cd].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results