accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNOSOO

CAS RN 35812-01-2

IUPAC Name: 2-bromo-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Synonyms: 2-bromo-1λ⁶,2-benzothiazole-1,1,3-trione
Molecular Weight (g/mol): 262.08
Molecular Formula: C7H4BrNO3S
InChi Key: QRADPXNAURXMSB-UHFFFAOYSA-N
SMILES: BrN1C(=O)C2=CC=CC=C2S1(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)