accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3582-71-6

IUPAC Name: 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane
Molecular Weight (g/mol): 277.36
Molecular Formula: C6H18Cl2O2Si3
InChi Key: GJIYNWRLGOMDEX-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)O[Si](C)(C)O[Si](C)(C)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results