accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35826-59-6

IUPAC Name: methyl 4-(propan-2-yloxy)benzoate
Synonyms: methyl 4-isopropoxybenzoate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: ROEMZKPINYTSKV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results