accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3584-23-4

IUPAC Name: 2-(4-methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Synonyms: 2-(4-methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 421.91
Molecular Formula: C12H7Cl6N3O
InChi Key: QRHHZFRCJDAUNA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=NC(=NC(=N1)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results