CAS RN 3584-66-5
IUPAC Name:
6-chloro-2-(trichloromethyl)-1H-1,3-benzodiazole
Synonyms:
5-chloro-2-(trichloromethyl)-3H-1,3-benzodiazole
Molecular Weight (g/mol):
269.93
Molecular Formula:
C8H4Cl4N2
InChi Key:
SIZGSKQSWJIWFP-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2N=C(NC2=C1)C(Cl)(Cl)Cl
1
–
1
of
1
results
1
Promotions Available
5-Chloro-2-(trichloromethyl)benzimidazole, 95%, Thermo Scientific™
5-Chloro-2-(trichloromethyl)benzimidazole, 95%, Thermo Scientific™
CAS: 3584-66-5 Molecular Formula: C8H4Cl4N2 Molecular Weight (g/mol): 269.93 MDL Number: MFCD00005595 InChI Key: SIZGSKQSWJIWFP-UHFFFAOYSA-N Synonym: 5-chloro-2-trichloromethyl benzimidazole,5-chloro-2-trichloromethyl-1h-benzo d imidazole,5-chloro-2-trichloromethyl-1h-benzimidazole,6-chloro-2-trichloromethyl-1h-benzimidazole,5-chloro-2-trichloromethyl-1h-1,3-benzodiazole,5-chloro-2-trichloromethyl-3h-1,3-benzodiazole,acmc-20am4z,5-chloro-2-trichloromethyl-1h-benzoimidazole PubChem CID: 77134 IUPAC Name: 6-chloro-2-(trichloromethyl)-1H-benzimidazole SMILES: ClC1=CC=C2N=C(NC2=C1)C(Cl)(Cl)Cl