accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOOCl

CAS RN 35856-62-3

IUPAC Name: piperidine-1-sulfonyl chloride
Synonyms: piperidine-1-sulfonyl chloride
Molecular Weight (g/mol): 183.65
Molecular Formula: C5H10ClNO2S
InChi Key: QQJYAXDCMMXECR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)N1CCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)