accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2O

CAS RN 3586-12-7

IUPAC Name: 3-phenoxyaniline
Synonyms: benzenamine, 3-phenoxy-
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: UCSYVYFGMFODMY-UHFFFAOYSA-N
SMILES: NC1=CC=CC(OC2=CC=CC=C2)=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results