accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOOHOHOOOO

CAS RN 358681-61-5

IUPAC Name: 5-(benzyloxy)-2-[(benzyloxy)methyl]-6-{[4,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-3-yl]oxy}oxane-3,4-diol
Synonyms: 5-(benzyloxy)-2-[(benzyloxy)methyl]-6-{[4,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-3-yl]oxy}oxane-3,4-diol
Molecular Weight (g/mol): 899.05
Molecular Formula: C54H58O12
InChi Key: XEEOIRLVLCPQPN-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(OC3OC(COCC4=CC=CC=C4)C(O)C(O)C3OCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C2OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results