CAS RN 3588-60-1
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
Synonyms:
2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
Molecular Weight (g/mol):
265.31
Molecular Formula:
C14H19NO4
InChi Key:
USPFMEKVPDBMCG-UHFFFAOYNA-N
SMILES:
CC(C)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Carbobenzoxy-DL-leucine 98.0+%, TCI America™
CAS: 3588-60-1 Molecular Formula: C14H19NO4 Molecular Weight (g/mol): 265.309 MDL Number: MFCD00065129 InChI Key: USPFMEKVPDBMCG-UHFFFAOYSA-N Synonym: N-Cbz-DL-leucine, Z-DL-Leu-OH PubChem CID: 246673 IUPAC Name: 4-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid SMILES: CC(C)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1